Description
For use with Diaphragm Vacuum Pumps only
Provides manual control and monitoring of vacuum levels. Analog gauge shows vacuum from 0–30 in.Hg. At the end of concentration, the control valve may opened to break the vacuum.
Specifications
Specifications
| For Use With | Diaphragm vacuum pump |
| Includes | Analog gauge showing 0 to 30 in. Hg and rear inlet and outlet ports |
| Type | Vacuum Controller |