Learn More
Labconco™ Vacuum Controller for CentriVap™ Systems
Product Code. 15123105 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With:
Diaphragm vacuum pump
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
15123105 Diaphragm vacuum pump Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15123105 Supplier Labconco™ Supplier No. 7991000
Only null left
Add to basket
Add to basket
For use with Diaphragm Vacuum Pumps only
Provides manual control and monitoring of vacuum levels. Analog gauge shows vacuum from 0–30 in.Hg. At the end of concentration, the control valve may opened to break the vacuum.
Specifications
For Use With Diaphragm vacuum pump
Includes Analog gauge showing 0 to 30 in. Hg and rear inlet and outlet ports
Type Vacuum Controller