Learn More
Labconco™ Tubing Kit for FreeZone freeze dry systems
Shop All Labconco Corporation Products
Click to view available options
Diameter (Metric) Inner:
1.95 cm
Unit Size:
Each
Description
3 feet of clear, reinforced vacuum hose for use with FreeZone Freeze Dryers.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Diameter (English) Inner | 3/4 in. |
| Diameter (Metric) Inner | 1.95 cm |
| Material | PVC |
| Overall Length (English) | 3 ft. |
| Overall Length (Metric) | 0.914 m |
| For Use With | FreeZone™ freeze dry systems |
| Color | Clear |
| Product Type | Tubing |