Learn More
Labconco™ Secondary Trap Canister and Inserts for CentriVap™ System
Shop All Labconco Corporation Products
Click to view available options
Type:
Acid Trap Insert
Moisture Trap Insert
Radioisotope Trap Insert
Secondary Trap Canister
Solvent Trap Insert
Weight (Metric):
1.8 kg
2.3 kg
Unit Size:
Each
Description
Help to provide secondary protection to vacuum pumps in evaporation systems
- Clear polypropylene canister with mounting bracket, 17 in. (43cm) PVC tubing, and two clamps
- Canister accommodates trap inserts listed in the table
Specifications
Specifications
| Material | Polypropylene |
| For Use With (Equipment) | CentriVap™ system |
| Type | Solvent Trap Insert |
| Weight (English) | 4 lb. |
| Weight (Metric) | 1.8 kg |