Learn More
Labconco™ FreeZone™ Stoppering or Bulk Tray Dryer Accessories
Shop All Labconco Corporation Products
Click to view available options
Length (English):
7 in.
12 in.
13 in.
Weight (English):
3 lb.
5 lb.
Unit Size:
Each
Description
Accessories for FreeZone Stoppering and Bulk Tray Dryers.
Support Grid Stainless steel with removable plastic grid Provides support for stoppering ampules and other small containers Holds 144-12mm dia. containers Use with Labconco Stoppering Tray and Bulk Tray Dryers Tray with Slide-out Bottom Stainless steel with separate bottom that slides out to allow glassware containers direct contact with shelf Use with Labconco Stoppering Tray and Bulk Tray Dryers Shelf Spacers Stainless steel Place on one or two emply shelves to assist stoppering of partial loads of small vials or bottles
Specifications
Specifications
| Type | Tray |
| No. of Shelves | 0 |
| Material | Stainless Steel |
| For Use With (Equipment) | Freezone™ stoppering tray dryer |
| Includes | Slide out bottom |
| Width (English) | 12 in. |
| Width (Metric) | 30.5 cm |
| Dimensions (L x W) | 12 x 12 in. (35.6 x 30.5cm) |
| Length (English) | 12 in. |
| Length (Metric) | 35.6 cm |
| Show More |