Learn More
Labconco Digital Electronic Vacuum Gauge
Shop All Labconco Corporation Products
Click to view available options
Diameter (Metric) Outside:
12.7 mm
Unit Size:
Each
Description
Digital Electronic Vacuum Gauges provide an LCD display of vacuum measurement in mBar, PA or Torr. Range is 0.02 mBar to 5 mBar. Stainless steel casing. Includes a cord with 3/8 in. to 1/2 in. OD adapter for connection to either vacuum tubing or a valve on a drying chamber. All FreeZone Freeze Dryers include a digital vacuum gauge. The Dry Ice Benchtop Freeze Dryer or other freeze dryers may use the Digital Electronic Vacuum Gauge as an accessory.
Specifications
Specifications
| Diameter (Metric) Outside | 12.7 mm |
| Type | Digital Vacuum Gauge |
| Weight (English) | 10.0 lb. |
| Weight (Metric) | 4.5 kg |
| Voltage | 230 V |
| Case Material | Stainless Steel |