Learn More
Labconco™ Anti-Static Ionizer Fan
Product Code. p-4366684 Shop All Labconco Corporation Products
Click to view available options
Voltage:
230 V
Unit Size:
Each
Product Code. 15538723
Brand: Labconco™ 5234401
Circulate ionized air inside Labconco Glove Boxes.
Anti-Static Ionizer Fan circulates ionized air inside Labconco Protector and Precise Glove Boxes and XPert Weigh Boxes to reduce static charge. Fan requires installation.
Specifications
For Use With (Equipment) Protector Combination Glove Box
Weight (English) 3.5 lb.
Weight (Metric) 1.6 Kg
Voltage 230 V
For Use With (Application) Circulates ionized air inside Glove Box to reduce Static Charge