missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ UltraVision™ DAB Away™
Shop All Epredia Products
Click to view available options
Volume (Metric):
250 mL
Unit Size:
250mL
Description
Includes:
- DAB Away Reagent 1 (125mL)
- DAB Away Reagent 2 (125mL)
- DAB Away Decolorizer (250mL)
Specifications
Specifications
| Content And Storage | Room temperature. Protect from light. |
| Volume (Metric) | 250 mL |
Safety and Handling
Product Identifier
- 250 ML ULTRAVISION DAB AWAY, INCLUDES DAB AWAY Reagent 1 (125mL), DAB Away Reagent 2 (125mL),
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- EUH210-Safety data sheet available on request.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P321-Specific treatment (see on this label).
- P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- MIXTURE LIST-Contains : Ethylene glycol, Orange G, Phosphotungstic acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction