Learn More
Epredia™ Lab Vision™ S100 Protein Ab-2 Rabbit Polyclonal Antibody with BSA and azide
Epredia™ S100 Protein Ab-2, Rabbit Polyclonal Antibody provides accurate, reproducible results.
Brand: Epredia™ RB-044-A0
Description
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an α and β chain whereas S100B is composed of two β chains. Antibody to S100 stains Schwannomas, ependymomas, astrogliomas, almost all benign and malignant melanomas and their metastases. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Cow, Kangaroo, Mouse, Rat and Chicken. Others not tested.
Epitope: Not determined
Immunogen: S100 protein purified from cow brain
Molecular Weight: ∽22kDa (nonreduced); ∽11kDa (reduced)
Positive Control: Melanoma or Schwannoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunocytochemistry
Note:
USA: RUO; Int'l: RUO
Specifications
| S100 Protein Ab-2 | |
| Polyclonal | |
| Unconjugated | |
| S100 protein purified from cow brain | |
| 100 μL | |
| Cancer and Tumor Biology | |
| Bovine, Chicken, Human, Kangaroo, Mouse | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Polyclonal | |
| Rabbit | |
| Melanoma or Schwannoma | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.