missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Nucleophosmin (NPM) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Take advantage of a quality product that delivers accurate results with the Epredia™ Nucleophosmin (NPM) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1849-PABX

 View more versions of this product

Product Code. 12690297

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Nucleophosmin (NPM)/B23/numatrin is a phosphoprotein associated with cell nucleoli. Treatment of cells with cytotoxic drugs may make nucleophosmin enter nucleoplasm. It is reported that nucleophosmin shuttles continuously between cytoplasm and nucleoli to carry newly synthesized ribosomal proteins into nucleoli. NPM-ALK is a hybrid of the anaplastic lymphoma kinase (ALK) gene and the nucleophosmin (NPM) gene resulting from the t(2;5)(p23;q35) translocation found in anaplastic large cell lymphoma. In acute promyelocytic leukemia NPM gene is fused to RARalpha gene in (5;17) (q33;q12) transition.

Host Species: Mouse

Clone: NA24

Isotype: IgG/κ

Species Reactivity: Human. Others not known.

Epitope: N-terminal

Immunogen: GST fusion protein containing the N-terminal part of nucleophosmin fused to 14aa of ALK protein

Molecular Weight: 38kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nucleophosmin (NPM) Ab-1
Monoclonal
1mg/mL
Mouse
200 μL
Cancer and Tumor Biology
Human
IgG κ
Immunohistochemistry (Paraffin)
NA24
Unconjugated
GST fusion protein containing the N-terminal part of nucleophosmin fused to 14 aa of ALK protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.