missing translation for 'onlineSavingsMsg'
Learn More

CD36/GPIIIb/GPIV Ab-3 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry , immunofluorescence, flow cytometry and function blocking experiment.

Brand:  Epredia MS-756-P0

 View more versions of this product

Product Code. 12602267

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

CD36 (also known as platelet glycoprotein IV, OKM5-antigen, or platelet glycoprotein IIIb) acts as a receptor for thrombospondin (TSP), collagen, falciparum malaria parasitized erythrocytes, and sickle erythrocytes. It also functions as a scavenger receptor, mediating macrophage uptake of oxidized low-density lipoprotein (LDL), and apoptotic polymorphonuclear leukocytes (PMN). CD36 plays a role in platelet aggregation, macrophage foam cell development, inflammation, and the tissue ischaemia observed in sickle cell disease and cerebral malaria. Ab-3 blocks adhesion of P. falciparum parasitized red blood cells to CD36 and strongly inhibits collagen-induced platelet aggregation.

Host Species: Mouse

Clone: 185-1G2

Isotype: IgG2a/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Stimulated human leukocytes

Molecular Weight: 88 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Frozen only)
  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Flow Cytometry
  • Function Blocking

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD36/GPIIIb/GPIV Ab-3
Monoclonal
200μg/mL
Mouse
100 μL
Hematopoietic
Human
IgG2a κ
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen)
185-1G2
Unconjugated
Stimulated human leukocytes
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.