Learn More
Eagle™ Lab Packs with Screw-On Lid
Product Code. 17742308
Change view
Click to view available options
Capacity (Metric):
75.7 L
113 L
217 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17742308 113 L Each
17752308 75.7 L Each
17762308 217 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17742308 Supplier Eagle™ Supplier No. EDR1600SYL
Only null left
Add to basket
Add to basket
Eagle Lab Packs are ideal for safe storage, handling and transportation of hazardous materials, spill containment and clean-up of hazardous materials, original shipping container, bio waste disposal and incineration.
  • Eagle Lab Pack Drums are constructed of Yellow blow-molded, high-density polyethylene (HDPE) with UV inhibitors, are durable and lightweight, weather and chemical resistant, and screw-on lid
  • Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
  • Lab Packs/Open Head Drums- for use with smaller non-damaged bottles, cans, carboys and 5-gallon pails of non-leaking hazardous materials - if leaking, must pack with sufficient sorbents for - NO FREE LIQUIDS
Specifications
Capacity (Metric) 113 L
Color Yellow
For Use With (Application) Safe Storage, Handling and Transportation of Hazardous Materials
Material Polyethylene