Learn More
L-(-)-threo-3-Hydroxyaspartic acid, TRC
Specifications
Specifications
| Chemical Name or Material | L-(-)-threo-3-Hydroxyaspartic acid |
| CAS | 7298-99-9 |
| Molecular Formula | C4 H7 N O5 |
| Quantity | 1 mg |
| InChI Formula | InChI=1S/C4H7NO5/c5-1(3(7)8)2(6)4(9)10/h1-2,6H,5H2,(H,7,8)(H,9,10)/t1-,2-/m0/s1 |
| SMILES | N[C@@H]([C@H](O)C(=O)O)C(=O)O |
| IUPAC Name | (2S,3S)-2-amino-3-hydroxybutanedioic acid |
| Molecular Weight (g/mol) | 149.102 |
| Formula Weight | 149.032 |