Learn More
Masterflex™ L/S™ Standard Pump Head Replacement Rotor Assembly
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Description
Masterflex™ L/S™ Standard Pump Head Replacement Rotor Assembly extends the operating life of standard pump heads
- Replacement parts extend the operating life of standard pump heads
- Two end bell assemblies are required for a complete head assembly
- Choose replacement parts suitable for your I/P™ standard pump head size
- Suitable for use with L/S™ Standard Pump Heads 07015-21, 07015-52, 07024-21 and 07024-52
Specifications
Specifications
| For Use With | L/S Standard Pump Heads 07015-21, 07015-52, 07024-21, and 07024-52 |
| Material | Stainless Steel |
| Tubing Type | High-Performance Precision Tubing |
| Type | Replacement Rotor Assembly |