Learn More
Masterflex™ L/S™ Replacement Motor for Digital Benchtop Modular Drive
Description
- Make the most of your Masterflex drives replacement parts and other accessories
- Extend the life of your Masterflex pump with these replacement parts
- Suitable for use with 77301-40 Benchtop Digital Modular Drive
Specifications
Specifications
| For Use With (Application) | Motor Replacement |
| For Use With | Benchtop Digital Modular Drive (77301-40) |
| Speed | 600 rpm |