Learn More
Masterflex™ L/S™ 2-Stop Precision Pump Tubing
Product Code. 11866743
Click to view available options
Tubing Size:
L/S 25
Unit Size:
Pack of 8
Product Code. 11866743
Brand: Masterflex™ 0642125
Only null left
Add to basket
Add to basket
Masterflex™ L/S™ 2-Stop Precision Pump Tubing with excellent biocompatibility that meets multiple classifications
  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance and pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match all unique applications
  • Always in stock - most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Slightly greater clarity than peroxide-cured tubing
  • Smooth surfaces; lower protein binding levels
  • Fewer potential leachables
  • Tubing Size: L/S 25
Specifications
Material Platinum-cured Silicone
Tubing Size L/S 25
Flow Rate ≤ 1000 mL/min
Hose Barb 3/16 in.
Includes 8/Pk.
Vacuum Level 660 mm Hg