Learn More
L-Malic Acid Disodium Salt Monohydrate, TRC
Product Code. 30480241
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
1g
2.50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480241 2.5 g 2.50g
30474743 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30480241 Supplier Toronto Research Chemicals Supplier No. TRCL2455952.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 207511-06-6
Molecular Formula C4H4O5Na2 . H2O
Molecular Weight (g/mol) 196.07
Synonym Butanedioic acid, 2-hydroxy-, sodium salt, hydrate (1:2:1), (2S)-, Butanedioic acid, hydroxy-, disodium salt, monohydrate, (2S)- (9CI), Disodium L-malate monohydrate, L-Malic acid disodium salt monohydrate
IUPAC Name disodium;(2S)-2-hydroxybutanedioate;hydrate
SMILES O.[Na+].[Na+].O[C@@H](CC(=O)[O-])C(=O)[O-]
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C4H6O5.2Na.H2O/c5-2(4(8)9)1-3(6)7;;;/h2,5H,1H2,(H,6,7)(H,8,9);;;1H2/q;2*+1;/p-2/t2-;;;/m0.../s1
Formula Weight 195.996
Chemical Name or Material L-Malic Acid Disodium Salt Monohydrate