missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-carnitine hydrochloride, 99%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Specifications
Specifications
| Color | White |
| Quantity | 1 g |
| Formula Weight | 197.7 |
| Percent Purity | 0.99 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-carnitine hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction