Learn More
L-beta-Proline ethyl ester hydrochloride, 97%, Thermo Scientific Chemicals
Product Code. 15404208
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15404208
Brand: Thermo Scientific Alfa Aesar H57781.03
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Protect from heat. Store away from oxidizing agents. Refrigerate. Incompatible with heat and oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
Molecular Formula C7H13NO2·HCl
MDL Number MFCD21337911
Synonym (S)-(+)-Pyrrolidine-3-carboxylic acid ethyl ester hydrochloride; Ethyl (S)-(+)-pyrrolidine-3-carboxylate hydrochloride
Specifications
Quantity 1 g
Formula Weight 179.65
Optical Rotation +14° (c=0.6 in Water)
Percent Purity 97%
Chemical Name or Material L-beta-Proline ethyl ester hydrochloride