missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartate de magnesium salt, MP Biomedicals™

Product Code. 11483251 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
1 kg
Unit Size:
1kg
25g
This item is not returnable. View return policy

Product Code. 11483251

Brand: MP Biomedicals 0215040625

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 18962-61-3
Molecular Formula C8H10MgN2O8-2
Molecular Weight (g/mol) 286.479
InChI Key XBYSRGUYNYEVEJ-UHFFFAOYSA-L
Synonym l-aspartic acid magnesium salt
PubChem CID 131850428
IUPAC Name magnesium;2-azanidylbutanedioate;hydron
SMILES [H+].[H+].C(C(C(=O)[O-])[NH-])C(=O)[O-].C(C(C(=O)[O-])[NH-])C(=O)[O-].[Mg+2]

Specifications

Color White
Quantity 25 g
Formula Weight 288.5 (Anhydrous)
Physical Form Crystalline Powder
Chemical Name or Material L-Aspartate de magnesium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.