missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartate de magnesium salt, MP Biomedicals™

Product Code. p-4404658 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
1 kg
Unit Size:
1kg
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483251 25 g 25g
11493251 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483251 Supplier MP Biomedicals Supplier No. 0215040625

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 18962-61-3
Molecular Formula C8H10MgN2O8-2
Molecular Weight (g/mol) 286.479
InChI Key XBYSRGUYNYEVEJ-UHFFFAOYSA-L
Synonym l-aspartic acid magnesium salt
PubChem CID 131850428
IUPAC Name magnesium;2-azanidylbutanedioate;hydron
SMILES [H+].[H+].C(C(C(=O)[O-])[NH-])C(=O)[O-].C(C(C(=O)[O-])[NH-])C(=O)[O-].[Mg+2]

Specifications

Color White
Quantity 25 g
Formula Weight 288.5 (Anhydrous)
Physical Form Crystalline Powder
Chemical Name or Material L-Aspartate de magnesium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.