Learn More
Kynurenic Acid, TRC
Product Code. 30003294
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003294 10 mg 10mg
30005831 500 mg 500mg
30004901 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30003294 Supplier Toronto Research Chemicals Supplier No. TRCK66050010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 492-27-3
Molecular Formula C10 H7 N O3
Molecular Weight (g/mol) 189.17
Synonym 2-Quinolinecarboxylic acid, 4-hydroxy- (9CI, ACI), 4-Hydroxy-2-quinolinecarboxylic acid (ACI), Quinaldic acid, 4-hydroxy- (8CI), 2-Carboxy-4-hydroxyquinoline, 4-Hydroxyquinaldic acid, 4-Hydroxyquinaldinic acid, 4-Hydroxyquinoline-2-carboxylic acid, Kynurenic acid, MeSH ID: D007736, NSC 58973, Quinurenic acid
IUPAC Name 4-hydroxyquinoline-2-carboxylic acid
SMILES OC(=O)c1cc(O)c2ccccc2n1
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C10H7NO3/c12-9-5-8(10(13)14)11-7-4-2-1-3-6(7)9/h1-5H,(H,11,12)(H,13,14)
Formula Weight 189.0426
Percent Purity >95%
Chemical Name or Material Kynurenic Acid
Grade HPLC