missing translation for 'onlineSavingsMsg'
Learn More

KW-2449, MedChemExpress

Product Code. p-200097852
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490207

Brand: Medchem Express HY10339

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KW-2449 is a multi-targeted kinase inhibitor of FLT3, ABL, ABLT315I and Aurora kinase with IC50s of 6.6, 14, 4 and 48 nM, respectively.

KW-2449 is a multi-targeted kinase inhibitor of FLT3, ABL, ABLT315I and Aurora kinase with IC50s of 6.6, 14, 4 and 48 nM, respectively.

Specifications

Chemical Name or Material KW-2449
Physical Form Powder
CAS 1000669-72-6
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H20N4O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 50 mg/mL (150.42 mM)
SMILES O=C(C1=CC=C(/C=C/C2=NNC3=C2C=CC=C3)C=C1)N4CCNCC4
Molecular Weight (g/mol) 332.4
Formula Weight 332.4
Percent Purity 99.72%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.