Learn More
Heidolph™ Kugelmeiers 3D Cell Culture Plate SP5D (Sphericalplate 5D)
Product Code. 17818808 Shop All Heidolph Instruments Products
Change view
Click to view available options
Array:
2 x 3
4 x 6
No. of Wells:
6
24
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Array No. of Wells unitSize
17818808 2 x 3 6 Each
17808808 4 x 6 24 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17818808 Supplier Heidolph™ Supplier No. 5491601000
The spherical plate 5D (SP5D) is a 3D cell culture plate for the formation of uniform, standardized spheroids with high quality and yield. It allows the effortless scaling and translation into the diagnostic or clinical application.
The patented geometry with special coating allows the integration of every single cell into a 3D-agglomerate in a controlled way. This is how uniformly sized spheroids grow without the formation of necrotic cores. The reproducibility of follow-up tests increases, since the experiments always start under identical initial conditions due to the low size variance of the cultivated spheroids.
  • According to the size and scale of the experiments, you can choose between our 24-well plate or our 6-well plate
  • For small batches, Heidolph™ suggests using the 24-well plate containing 12 wells with 750 microwells each so you will be able to yield up to 9000 uniform spheroids
  • You can comfortably carry out monolayer comparative studies in the remaining 12 wells of the same plate
  • 6-well plate is suitable for scale-up trials and bigger batches; you can expect a yield of 3364 spheroids per well, thus a total yield of 20,184 spheroids/plate
  • The ready to use SP5D is particularly user-friendly: The cultivation does not require any pretreatment or centrifugation steps after seeding your cells
  • The media change is also particularly convenient due to simple pipetting, since the height of the microwells was chosen so that the cell clusters are retained
  • Platform is compatible with standard automation units
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Lid With Lid
Array 2 x 3
Autoclavable Not Autoclavable
Color Transparent
Disposable Single-use
For Use With (Equipment) Standard Automation Unit
Material Cycloolefin Copolymer
No. of Wells 6
Sterility Sterile
Surface Treatment Non-binding
Well Shape Round, Conical Bottom (V-well)
For Use With (Application) Spheroid Cultivation
Includes Plate
Fit for Purpose Sterile and PCR Ready