missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT84 Monoclonal Antibody (OTI2D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811189S
This item is not returnable.
View return policy
Description
KRT84 Monoclonal antibody specifically detects KRT84 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| KRT84 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9HCI7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55167 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI2D1 | |
| Unconjugated | |
| MSL2 | |
| 65 kDa type II keratin; AV235125; hard keratin, type II, 4; HB4; HRb-1; K84; Kb24; keratin 84; keratin 84, type II; keratin complex 2, basic, gene 16; keratin complex 2, gene 16; keratin, hair, basic, 4; keratin, type II cuticular Hb4; Keratin-84; Krt2-16; Krt2-3; Krt84; KRTHB4; type II hair keratin 4; type II hair keratin Hb4; type II keratin Kb24; type-II keratin Kb24 | |
| Full length human recombinant protein of human MSL2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction