missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT14 Monoclonal Antibody (OTI4A7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803267S
This item is not returnable.
View return policy
Description
KRT14 Monoclonal antibody specifically detects KRT14 in Human samples. It is validated for Western BlotSpecifications
| KRT14 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P04275 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7450 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI4A7 | |
| Unconjugated | |
| VWF | |
| AI626930; CK14; CK-14; cytokeratin 14; cytokeratin-14; EBS3; EBS4; epidermolysis bullosa simplex, Dowling-Meara, Koebner; K14; Ka14; keratin 14; keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner); keratin 14, type I; keratin complex 1, acidic, gene 14; keratin, type I cytoskeletal 14; keratin, type I cytoskeletal 17; keratin-14; Koebner; Krt-1.14; Krt1-14; KRT14; LOC100737113; NFJ; Type I keratin Ka14 | |
| Human recombinant protein fragment corresponding to amino acids 764-1263 of human VWF produced in SF9 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction