Learn More
KRN-633, MedChemExpress
Product Code. 16491207
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16491207
Brand: Medchem Express HY12060
MedChemExpress KRN-633 is a potent VEGFR inhibitor with IC50s of 170, 160 and 125 nM for VEGFR1, VEGFR2 and VEGFR3, respectively.
KRN-633 is a potent VEGFR inhibitor with IC50s of 170, 160 and 125 nM for VEGFR1, VEGFR2 and VEGFR3, respectively.
Specifications
Chemical Name or Material KRN-633
CAS 286370-15-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H21ClN4O4
Quantity 50 mg
Solubility Information DMSO : 25 mg/mL (59.97 mM; Need ultrasonic)
SMILES ClC1=CC(OC2=NC=NC3=CC(OC)=C(C=C32)OC)=CC=C1NC(NCCC)=O
Molecular Weight (g/mol) 416.86
Formula Weight 416.86
Percent Purity 99.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease