Learn More
Diversey™ Kristalle VB79 Acidic Detergent Synergist
Specifically designed for packaging aseptic. Diversey™ Kristalle VB79 Acidic Detergent Synergist decreases the surface tension of solutions of peracetic acid (APA) to promote contact between the disinfectant and packaging plastics that are highly hydrophobic in nature.
Brand:  Diversey™ 7511668
Product Code. 15379009
  • ONLINE EXCLUSIVE PRICE
    311.00 EUR
    252.77 EUR / 20 L
    Save 58.23 EUR
    18% Off
Description
Key Properties
  • Decreases the surface tension of solutions of peracetic acid (APA) to promote contact between the disinfectant and packaging plastics that are highly hydrophobic in nature; in this way the action disinfectant is maximized without the need for excessive concentrations
  • Promotes the dewatering and recovery of disinfectant solutions, thus contributing to a lower cost of disinfection
  • Can be determined in proportion to the contributions of APA
  • Recommended for pharmaceutical and cosmetic industries
  • Also used in food industry, alone, or in CIP additive for detergent sufactant properties, on difficult stains or brittle materials
Advantages
  • Strong wetting properties on plastic
  • Contributes to the action of the disinfectant by promoting contact with the surface
  • No action on the stability of the APA
  • Compatible with all water hardnesses
  • Low foaming
Use and Storage
  • Use proportional doses of APA refills and to concentrations between 0.02 and 1% v/v depending on the type of support and selected temperature
  • Must be completely rinsed with water after application to remove any residue on the surfaces in contact with foodstuffs
  • Store in original closed container away from extreme temperatures
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Kristalle VB79
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contains : alkyl alcohol alkoxylate