Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with PTFE-Faced Fritted Glass Support (47mm)
Product Code. 11838502
Change view
Click to view available options
Volume (Metric):
300 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11838502 300 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11838502 Supplier DWK Life Sciences Supplier No. 9537800000
Used for autoclaving with the filter in place and is designed to handle up to 500mL of sample liquids for the analysis of particulate or microbiological contamination
  • Supplied with a 40-60 micron porosity PTFE-faced fritted glass support base, a 300mL PTFE-faced graduated funnel, an anodized aluminum clamp and a #8 silicone stopper
  • PTFE coating prevents membrane from adhering to ground glass surface
  • Funnels are graduated from 100 to 250mL in 25mL increments
  • Prefilter size is 35mm
  • Approximate filter area is 9.6cm2
  • Connection to 1 liter filtration flask (available as an accessory) is made with a #8 silicone stopper
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE
Volume (Metric) 300 mL
Width (English) 3 in.
Width (Metric) 7.6 cm
Height (English) 9 in.
Height (Metric) 22.9 cm