Learn More
Kontes™ Membrane Filtration Glassware from Restek
Product Code. 11848522
Change view
Click to view available options
Type:
Flask (2 L)
Flask (4 L)
Funnel (1 L)
Funnel (300 mL)
Funnel (500 mL)
Support
Material:
Fritted Glass
Glass
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Material unitSize
11848522 Funnel (1 L) Glass Each
11739055 Support Fritted Glass Each
11888512 Flask (2 L) Glass Each
11828502 Funnel (500 mL) Glass Each
11818522 Funnel (300 mL) Glass Each
11729055 Flask (4 L) Glass Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11848522 Supplier Kimble Chase Supplier No. 9537810000
Used for routine filtration of corrosive liquids and removal of particles from HPLC solvents
  • Ground joint connection eliminates phthalate contamination that can occur when using silicone or neoprene stoppers
  • Support base has a coarse porosity glass frit and an integral vacuum connection, located above the drip tip that prevents contamination of the vacuum line with filtrate droplets
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Microfiltration Apparatus
Material Glass
Type Funnel (1 L)
Volume (Metric) 1 L
Length (Metric) 47 mm