Learn More
DWK Life Sciences Kimble™ DISPOSAFLEX™ Polypropylene Disposable Chromatography Funnels Only
Click to view available options
Unit Size:
Pack of 100
Description
- 9mL Polypropylene Reservoir for use with DISPOSAFLEX™ Columns
- Case of 100
Specifications
Specifications
| For Use With (Equipment) | Disposaflex™ disposable chromatography funnel |
| Type | Reservoir |
| Column Capacity | 9 mL |
| Material | Polypropylene |