missing translation for 'onlineSavingsMsg'
Learn More

Kobe0065, MedChemExpress

Product Code. 16431196
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16431196

Brand: Medchem Express HY15716

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Kobe0065 is a novel and effective inhibitor of Ras-Raf interaction, competitively inhibiting the binding of H-Ras·GTP to c-Raf-1 RBD with a Ki value of 46±13 μM.

Kobe0065 is a novel and effective inhibitor of Ras-Raf interaction, competitively inhibiting the binding of H-Ras·GTP to c-Raf-1 RBD with a Ki value of 46±13 μM.

Specifications

Chemical Name or Material Kobe0065
Physical Form Solid
CAS 436133-68-5
Color Yellow
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C15H11ClF3N5O4S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 42.5 mg/mL (94.49 mM)
SMILES FC(C1=CC([N+]([O-])=O)=C(NNC(NC2=CC=C(C)C(Cl)=C2)=S)C([N+]([O-])=O)=C1)(F)F
Molecular Weight (g/mol) 449.79
Formula Weight 449.79
Percent Purity 98.01%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.