missing translation for 'onlineSavingsMsg'
Learn More

KLK8 Monoclonal Antibody (OTI10F9), TrueMAB™, OriGene

Product Code. 17565582
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17565582 30 μL 30µL
17545582 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17565582 Supplier Origene Technologies Supplier No. TA506162S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KLK8 Monoclonal antibody specifically detects KLK8 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen KLK8
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI10F9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PTK2
Gene Accession No. Q05397
Gene Alias brain serine protease 1; BSP1; EGK_10950; H NRPN; hK8; HNP; kallikrein 8; kallikrein 8 (neuropsin/ovasin); kallikrein related peptidase 8; kallikrein related-peptidase 8; Kallikrein8; kallikrein-8; kallikrein-related peptidase 8; Klk8; mK8; neuropsin; NP; Nrpn; Ovasin; PRO322; protease, serine, 19 (neuropsin); Prss19; Serine protease 19; serine protease TADG-14; TADG14; Tumor-associated differentially expressed gene 14 protein; UNQ283; UNQ283/PRO322
Host Species Mouse
Immunogen Full length human recombinant protein of human PTK2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5747
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.