missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLHL3 Polyclonal Antibody

Description
The antibody detects endogenous levels of total KLHL3 protein.
Probable substrate-specific adapter of an E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
Specifications
Specifications
| Antigen | KLHL3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | KLHL3 |
| Gene Accession No. | E0CZ16, Q9UH77 |
| Gene Alias | 7530408C15Rik; AI430941; EG627648; kelch like family member 3; kelch-like 3; kelch-like family member 3; kelch-like protein 3; KIAA1129; Klhl3; PHA2D; RGD1565218 |
| Gene Symbols | KLHL3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?