Learn More
LNI Swissgas Kit Service for NG SIRIO 10 L
Product Code. 17832049 Shop All LNI Swissgas Products
Click to view available options
Type:
PM Kit
Unit Size:
Each
Product Code. 17832049
Brand: LNI Swissgas 6930.00.195
This item requires a price update prior to being ordered.
Request a Price for this item here.
Kit service 8000 h for NG SIRIO 10L.
  • To be carried out by trained service engineer every 16000 hours or 4 years.
Specifications
Type PM Kit