missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIR2D Monoclonal Antibody (NKVFS1)
GREENER_CHOICE

Mouse Monoclonal Antibody

552.00 EUR

Specifications

Antigen KIR2D
Clone NKVFS1
Concentration 1 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11560173
GREENER_CHOICE
View Documents
Invitrogen™
MA182651
100 μg
552.00 EUR
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15206119
View Documents
Invitrogen™
MA516887
200 μg
552.00 EUR
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Clone NKVFS1 recognizes the long and short forms CD158a and CD158b (KIR2DL, KIR2DS1 and KIR2DS2 respectively) and also p50.3 (KIR2DS4). This clone is reported to have functional activity, activating NK cell cytotoxicity via KIR2DS and inhibiting via KIR2DL forms. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

Killer cell immunoglobulin-like receptors (KIRs) are transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. The KIR genes are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC). The gene content of the KIR gene cluster varies among haplotypes, although several 'framework' genes are found in all haplotypes. The KIR proteins are classified by the number of extracellular immunoglobulin domains (2D or 3D) and by whether they have a long (L) or short (S) cytoplasmic domain. KIR proteins with the long cytoplasmic domain transduce inhibitory signals upon ligand binding via an immune tyrosine-based inhibitory motif (ITIM), while KIR proteins with the short cytoplasmic domain lack the ITIM motif and instead associate with the TYRO protein tyrosine kinase binding protein to transduce activating signals. The ligands for several KIR proteins are subsets of HLA class I molecules; thus, KIR proteins are thought to play an important role in regulation of the immune response.
Specifications

Specifications

KIR2D
1 mg/mL
ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Unconjugated
Mouse
RUO
PBS with 0.09% sodium azide; pH 7.4
P43626, P43631, P43632, Q14953, Q14954, Q99706
100132285, 3802, 3805, 3806, 3809, 3810
Proprietary Immunogen.
Antibody
NKVFS1
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Monoclonal
Liquid
IgG1
Human
KIR2DL1
183ActI; CD158; CD158 antigen-like family member A; CD158 antigen-like family member G; CD158 antigen-like family member H; CD158 antigen-like family member I; CD158 antigen-like family member J; CD158A; CD158b; CD158B1; CD158G; CD158H; CD158I; CD158j; cl-49; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 1; killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 1; killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 2; killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4; killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 5; killer cell immunoglobulin-like receptor 2DL1; Killer cell immunoglobulin-like receptor 2DS1; killer cell immunoglobulin-like receptor 2DS2; Killer cell immunoglobulin-like receptor 2DS4; killer cell immunoglobulin-like receptor 2DS5; killer cell immunoglobulin-like receptor KIR2DS2; killer cell immunoglobulin-like receptor KIRDS1; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 1; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 1; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 2; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 4; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5; killer Ig receptor; killer inhibitory receptor 2-2-1; killer inhibitory receptor 4-1-2; killer-cell Ig-like receptor; killer-cell immunoglobulin-like receptor; killer-cell immunoglobulin-like receptor two domains short tail 2 protein; KIR antigen 2DS4; KIR1D; KIR221; KIR2DL1; KIR2DP1DL1; KIR2DS1; KIR2DS2; KIR-2DS2; KIR2DS4; KIR-2DS4; KIR2DS5; KIR412; KIR-K64; KKA3; MHC class I NK cell receptor; MHC class I NK cell receptor Eb6 ActI; natural killer associated transcript 5; natural killer cell inhibitory receptor; natural killer cell receptor; natural killer-associated transcript 1; Natural killer-associated transcript 5; Natural killer-associated transcript 8; natural killer-associated transcript 9; NK receptor 183 ActI; NKAT; NKAT1; NKAT-1; NKAT5; NKAT-5; NKAT6; NKAT8; NKAT-8; NKAT9; NKAT-9; p50 killer cell activating receptor KAR-K1d; p50.1; p50.3; p58 killer cell inhibitory receptor KIR-K64; p58 killer cell inhibitory receptor KIR-K7a; p58 natural killer cell receptor clone CL-49; P58 natural killer cell receptor clones CL-39/CL-17; p58 natural killer cell receptor clones CL-42/47.11; p58 NK cell inhibitory receptor NKR-K6; p58 NK receptor CL-39/CL-17; p58 NK receptor CL-42/47.11; p58 NK receptor CL-49; p58.1; p58.1 MHC class-I-specific NK receptor; XXbac-BCX195L8.1
KIR2DL1, KIR2DL4, KIR2DS1, KIR2DS2, KIR2DS4, KIR2DS5
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.