Learn More
Thermo Scientific™ KingFisher Plastic Consumables for SARS-CoV-2 Testing
Shop All Thermo Scientific ProductsDescrizione
These KingFisher compatible plastic consumables are offered specifically for labs performing COVID-19 testing with the MagMAX Viral/Pathogen Nucleic Acid Purification Kit and the TaqPath COVID-19 Combo Kit.
For TaqPath COVID-19 testing with MagMAX Viral/Pathogen Nucleic Acid Isolation Kit
Specifica
Specifica
| Format | 96-well Plate |
| Capacity | 96 Samples |
| For Use With (Equipment) | KingFisher Flex and Presto |
| Includes | KingFisher DW 96 Tip Comb |
| Quantity | 100/Pk. |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.