missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab King B Agar (F Agar)
Click to view available options
:
King B Agar; Quantity: 500 g
Description
- Pigment production agar B
- Enhances fluorescein production in Pseudomonas species
- DIN 38411, ISO 16266, 22717 and EN 12780 standards
Specifications
Specifications
| Certifications/Compliance | DIN 38411, ISO 16266, 22717, EN 12780 |
| Form | Solid |
| Media Type | King B Agar |
| Product Type | Microbiology Media |
| Formulation | Meat Peptone 10.0 g/L, Casein Peptone 10.0 g/L, Dipotassium Phosphate 1.5 g/L, Magnesium Sulfate 1.5 g/L, Agar 15.0 g/L |
| For Use With (Application) | Cell Culture |
| Incubation Conditions | 30°C ± 1°C |
| Incubation Time | 44 ±4 hr. |
| pH | 7.2 ±0.2 |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction