missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kinesin 5B Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-580PCP
This item is not returnable.
View return policy
Description
Kinesin 5B Polyclonal antibody specifically detects Kinesin 5B in Human, Mouse, Porcine samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Kinesin 5B | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/ Immunofluorescence | |
| Conventional kinesin heavy chain, kinesin family member 5B, kinesin-1 heavy chain, KNS1kinesin 1 (110-120kD), KNSKINH, Ubiquitous kinesin heavy chain, UKHCkinesin heavy chain | |
| Stalk domain of human kinesin (amino acids 331-906) expressed in E. coli (FKHC3). | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience | |
| 3799 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| PerCP | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction