Learn More
Kinesin 5B Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB500-580H
Product Code. 18562221
  • 547.00 EUR / 0.10mL
Description
Kinesin 5B Polyclonal antibody specifically detects Kinesin 5B in Human, Mouse, Porcine samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Kinesin 5B
Polyclonal
PBS
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Pig
Purified
Western Blot
HRP
Conventional kinesin heavy chain, kinesin family member 5B, kinesin-1 heavy chain, KNS1kinesin 1 (110-120kD), KNSKINH, Ubiquitous kinesin heavy chain, UKHCkinesin heavy chain
Stalk domain of human kinesin (amino acids 331-906) expressed in E. coli (FKHC3).
0.1 mL
Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience
3799
Store at 4°C in the dark.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions