missing translation for 'onlineSavingsMsg'
Learn More

KIN1148, MedChemExpress

Product Code. 16474736
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16474736

Brand: Medchem Express HY101950

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KIN1148, a small-molecule IRF3 agonist, is a novel influenza vaccine adjuvant found to enhance flu vaccine efficacy.

KIN1148, a small-molecule IRF3 agonist, is a novel influenza vaccine adjuvant found to enhance flu vaccine efficacy.

Specifications

Chemical Name or Material KIN1148
Physical Form Solid
CAS 1428729-56-9
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H11N3OS2
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : ≥ 100 mg/mL (276.67 mM)
SMILES O=C(NC1=NC2=CC=C3C(N=CS3)=C2S1)C4=CC5=C(C=C4)C=CC=C5
Molecular Weight (g/mol) 361.44
Formula Weight 361.44
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.