Learn More
Kimberly-Clark™ Kimtech™ Pure W4 Wipers, 5 Packs x 100 White Cloths
Product Code. p-200080451 Shop All Kimberly-Clark Professional Products
Click to view available options
Dimensions (L x W):
30.4 x 30.4 cm
Unit Size:
Pack of 500
Product Code. 12685606
missing translation for 'mfr': Kimberly-Clark™ 7605
Only null left
Add to basket
Add to basket
Suitable for ISO Class 4 or higher cleanroom environments. Perfect for dry wiping critical equipment and surfaces in pharmaceutical, biotechnology, medical device and food processing industries. 30.4cm (W). Double bagged with case liner.
  • 5 packs x 100 white cloths (500 total)
  • Low linting, 100% Polypropylene white base sheets
  • Perfect for pharmaceutical, high-tech biotechnology, medical device and food processing industries
  • Suitable for ISO Class 4, or higher, cleanroom environments
  • Acid, base and solvent resistant
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Wiper
Length (English) 12.59 in.
Length (Metric) 32.7 cm
Width (English) 12.59 in
Width (Metric) 32.7 cm
Material Paper
Color White
Dimensions (L x W) 30.4 x 30.4 cm
Packaging Quantity 100 Sheets x 5 Bags (500 total)