Learn More
Shamrock™ Kimdura Durastick Label
Click to view available options
Dimensions (LxW):
11 mm
12 x 32 mm
Unit Size:
Pack of 1000
Specifications
Specifications
| Adhesive Type | Permanent |
| Color | White |
| Material | Polypropylene |
| Product Type | Durastick Label |
| For Use With (Application) | Cryogenic use |
| For Use With (Equipment) | Thermal printer |
| Min. Operating Temperature (Metric) | -80°C |
| Size (Metric) | 12 x 32 mm |
| Dimensions (LxW) | 12 x 32 mm |