Learn More
DWK Life Sciences Kimble™ Glass Screw-Thread Sample Vials without Closure
Product Code. p-205137
Click to view available options
Certifications/Compliance:
ASTM Type I, Class A
USP Type I
Color:
Amber
Clear
Closure Size:
15-425
18-400
Volume (English):
1 dr.
2.16 dr.
Unit Size:
Pack of 1000
Product Code. 17751078
Brand: DWK Life Sciences 608151760
Only null left
Add to basket
Add to basket
Cylindrical shape provides solid balance. Flat bottom facilitates free-standing capability.
  • Glass sample vial has a screw thread finish
  • KG-33 borosilicate glass
  • Ungraduated with no closure included
TRUSTED_SUSTAINABILITY
Specifications
Color Amber
Diameter (Metric) Outer 17 mm
Material Borosilicate Glass
Volume (Metric) 8.0 mL
Thread Style Screw
Volume (English) 2.16 dr.
Closure Type Screw Cap
Height (Metric) 60 mm
Certifications/Compliance USP Type I
Closure Size 15-425