Learn More
DWK Life Sciences Kimble™ Kontes™ NMR Pipets
Extended-tip NMR pipets for retrieving and adding sample in NMR tubes
218.00 EUR - 233.00 EUR
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11864065
|
DWK Life Sciences
897085-0009 |
NMR Pipet |
218.00 EUR
Pack of 100 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
11803153
|
DWK Life Sciences
897085-0007 |
Extended Tip Pipet |
233.00 EUR
Pack of 100 |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
Description
Extended-tip NMR pipets for retrieving and adding sample in NMR tubes
- Available with 7 and 9 in. (17.7 and 23cm) tips for use with 7 and 9 in. long NMR tubes
Specifications
Borosilicate Glass | |
100/Cs. |