Learn More
DWK Life Sciences Kimble™ Kontes™ NMR Pipets
Extended-tip NMR pipets for retrieving and adding sample in NMR tubes
Brand:  DWK Life Sciences 897085-0007
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11803153
  • 233.00 EUR / Pack of 100
Estimated Shipment: 05-08-2024
Description
Extended-tip NMR pipets for retrieving and adding sample in NMR tubes
  • Available with 7 and 9 in. (17.7 and 23cm) tips for use with 7 and 9 in. long NMR tubes
Specifications
Specifications
Borosilicate Glass
100/Cs.
Extended Tip Pipet