Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials without Closures
Product Code. 11852983
Click to view available options
Volume (Metric):
1.8 mL
2 mL
4 mL
6 mL
8 mL
12 mL
16 mL
20 mL
24 mL
25 mL
40 mL
Diameter (Metric) Outer:
12 mm
15 mm
17 mm
19 mm
21 mm
23 mm
28 mm
Closure Size:
13-425
15-425
18-400
20-400
24-400
8-425
Unit Size:
Pack of 200
Product Code. 11852983
Brand: DWK Life Sciences 608121545
Only null left
Add to basket
Add to basket
Packaged in corrugated trays with partitions
  • Manufactured from clear 33 expansion borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Diameter (Metric) Outer 15 mm
Material Borosilicate Glass
Volume (Metric) 4 mL
Thread Style Screw
Volume (English) 1 dr.
Height (Metric) 45 mm
Certifications/Compliance ASTM Type I, Class A
Closure Size 13-425