Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials without Closures
Click to view available options
Volume (Metric):
1.8 mL
2 mL
4 mL
6 mL
8 mL
12 mL
16 mL
20 mL
24 mL
25 mL
40 mL
Diameter (Metric) Outer:
12 mm
15 mm
17 mm
19 mm
21 mm
23 mm
28 mm
Closure Size:
13-425
15-425
18-400
20-400
24-400
8-425
Unit Size:
Pack of 200
Description
- Manufactured from clear 33 expansion borosilicate glass
Specifications
Specifications
| Color | Clear |
| Diameter (Metric) Outer | 12 mm |
| Material | Borosilicate Glass |
| Volume (Metric) | 1.8 mL |
| Thread Style | Screw |
| Volume (English) | 0.5 dr. |
| Height (Metric) | 32 mm |
| Certifications/Compliance | ASTM Type I, Class A |
| Closure Size | 8-425 |