Learn More
DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached Closures
Click to view available options
Volume (Metric):
20 mL
40 mL
Height (Metric):
57 mm
95 mm
Unit Size:
Pack of 72
Description
- Manufactured from clear 33 Expansion borosilicate glass
- Closures supplied with PTFE-faced silicone liners
- Vials are packaged in corrugated trays with partitions
Specifications
Specifications
| Classification | A |
| Material (Liner) | PTFE/Silicone |
| Color | Clear |
| Diameter (Metric) Outer | 28 mm |
| Material | Borosilicate Glass |
| Volume (Metric) | 40 mL |
| Thread Style | Screw |
| Volume (English) | 10.82 dr. |
| Closure Material | Polypropylene |
| Closure Type | Screw Cap (Open-top) |
| Show More |