Learn More
DWK Life Sciences Kimble™ Black Phenolic Screw-Thread Closures
Click to view available options
Neck Finish:
13-415
15-415
Unit Size:
Pack of 500
Description
- Cemented-in PTFE-faced 14B white rubber liner
- Three screw-thread finishes
Specifications
Specifications
| Color | Black |
| Material | Phenolic Resin |
| No. per Case | 500 Cs. |
| Neck Finish | 13-415 |
| Certifications/Compliance | ASTM E982 |
| Product Type | Screw Cap |
| For Use With (Equipment) | Bottle |
| Closure Liner | PTFE/14B Rubber |
| Autoclavable | Autoclavable |