Learn More
DWK Life Sciences Kimble™ Adapters for Round-Bottom Centrifuge Tubes
Click to view available options
Capacity:
15 mL Round-bottom Centrifuge Tube
30 mL Round-bottom Centrifuge Tube
Unit Size:
Pack of 2
Description
Required when centrifuging select Kimble high-strength round-bottom centrifuge tubes to 13,000xG in 50mL rotor
- Made of rubber
Specifications
Specifications
| Type | Centrifuge Tube Adapter |
| Material | Rubber |
| Capacity | 15 mL Round-bottom Centrifuge Tube |